Learn More
Chemglass Life Sciences Cap, Chem Snap, 11mm, Clear, 10mil PTFE Septum
Supplier: Chemglass Life Sciences CV35500011
Cap, Chem Snap, 11mm, Clear, 10mil PTFE Septum. Chem Snap Caps may be snapped on and off by hand which eliminates the need for metal crimping and decapping tools, thus reducing potential hazards due to sharp aluminum edges. Large opening provides greater target area for needle penetration. Snap caps are color coded for easy sample identification.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.