Learn More
Cell Systems Corporation Complete Medium Formulated at Normal Blood Glucose Level With Serum and CultureBoost (4N0-500)
Supplier: Cell Systems Corporation 4N0500
Our medium is designed with normal human blood glucose level and is formulated with 10% serum. It becomes a complete medium once activated with the included CultureBoost supplement (4CB-500). This kit consists of 500mL Normal Glucose Medium, 5mL CultureBoost (containing animal derived growth factors) and 10mL Attachment Factor (extracellular matrix reagent for coating flasks). 4N0-500 is certified and intended for experimental application. The media is phenol-red free with no added hormones or antibiotics for optimized experimental conditions. It is certified to be used with more than 100 human and animal cell cultures and cell lines. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Our products are available for next-day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.