Learn More
Cell Systems Corporation Medium Kit Without Serum and With CultureBoost (4Z3-500)
Supplier: Cell Systems Corporation 4Z3500
Our base medium without serum is customizable to meet your research-specific needs. For complete medium, add 5-20% serum (defined FBS or equivalent) (not included) and the included CultureBoost-R supplement. This kit consists of 500mL 4Z3-500 Medium, 5mL CultureBoost (containing animal derived growth factors) and 10mL Attachment Factor (extracellular matrix reagent for coating flasks). 4Z3-500 is certified and intended for experimental application. The media is phenol-red free with no added hormones or antibiotics for optimized experimental conditions. It is certified to be used with more than 100 human and animal cell cultures and cell lines. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Our products are available for next-day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.