Learn More
Thermo Scientific Dionex IonPac CS20 IC Columns | | 1 EA
Supplier:  Thermo Scientific 302608
Encompass
Analyze inorganic cations and amines using the Thermo Scientific Dionex IonPac CS20 ion chromatography IC columns. Designed to complement Dionex IonPac CS18 and Dionex IonPac CS19 columns, the Dionex IonPac CS20 columns provide a large separation window between monovalent and divalent cations. This allows most amines to elute after the alkali metals but before the alkaline earth metals, moving the amines away from matrix ions that would normally make quantitation difficult. Its selectivity makes the Dionex IonPac CS20 particularly useful for determining a variety of alkyl and alkanol amines.
Catalog No. NC1287363
Explore available promotions