Learn More
Med Vet International ProVetLogic ProFoam2 Multi-Dilution Gun
Supplier:  Med Vet International V97
Encompass_Preferred
ProFoam2 is a hose end 96 oz. foamer offering automatic dilution with convenient control of where and how ProVetLogic concentrated cleaning solutions are applied. Application of diluted solutions with the ProFoam2 hose-end proportioner is easy. On and off functions are finger-tip controlled using a trigger on the spray nozzle. ProFoam2 eliminates the time and hassle of manually mixing concentrates, which not only increases labor productivity, but also increases concentrate performance because the solutions are consistently mixed as directed by the manufacturer. Since concentrates are mixed only as they¿re applied, they are fresh and will perform at peak power, and waste is minimized.
Catalog No. 50-209-1759
Due to product restrictions, please to purchase or view availability for this product.
Explore available promotions