Learn More
Leap PAL Parts Headspace Magnetic Crimp Cap 20mm, 5mm hole, PTFE/Clear Silicone, 3mm, 100 Pack
Supplier:  Leap PAL Parts XAR20038385
Encompass_Preferred
Cap, Crimp, Headspace, Magnetic With 5mm hole, 0.01 in. PTFE/Clear Silicone - 2.5mm, 100 Pack
Catalog No. 50-635-116
Explore available promotions