Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Inserts for 1.5 mL Large Opening Vials
Allow the use of smaller sample volumes with standard autosampler vials.
Supplier: MilliporeSigma™ Supelco™ SU860067
Description
- Vial inserts for 1.5 mL large opening vials [0.1 mL, clear glass conical (with bottom spring)] allow the use of smaller sample volumes with standard autosampler vials.
- Inserts are generally chosen based on the inner diameter of the vial neck.
- Large opening vials (6 mm ID) require a 6 mm OD insert.
- Clear glass conical inserts used with CTC LC PAL autosamplers.
Specifications
4.6 mm | |
Conical Clear Glass | |
100 |
6 mm | |
0.1 mL | |
31 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction