Learn More
Kamiya Biomedical Liquid ITA Control, L1: 2 x 2mL, L2: 2 x 2mL RX
Supplier: Kamiya Biomedical K49C4M
K-ASSAY Liquid ITA Control. Level 1: 2 x 2 mL (liquid), Level 2: 2 x 2 mL (liquid). The K-ASSAY Liquid ITA Control is intended to be used as a consistent test sample of known concentration for monitoring the performance of K-ASSAY immunoturbidimetric assays (ITA) for the listed constituents: Alpha-1 Acid Glycoprotein (Cat. No. KAI-021), Alpha-1 Anti-Trypsin (Cat. No. KAI-001), C-Reative Protein (Cat. No. KAI-026), Complement C3 (Cat. No. KAI-009), Complement C4 (Cat. No. KAI-010), Haptoglobin (Cat. No. KAI-022), IgA (Cat. No. KAI-013), IgG (Cat. No. KAI-014), IgM (Cat. No. KAI-015), Prealbumin (Cat. No. KAI-053), and Transferrin (Cat. No. KAI-023). FOR IN VITRO DIAGNOSTIC USE.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.