Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEOX1 293T Cell Overexpression Lysate (Denatured), Abnova
Supplier: Abnova Corporation H00004222T01
Description
Western Blotting
Specifications
Western Blot | |
MEOX1 293T Cell Transient Overexpression Lysate(Denatured) | |
28.05 | |
pCMV-MEOX1 full-length | |
100 μL | |
Human |
293T | |
NM_004527.3 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-MEOX1 antibody (H00004222-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction