Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP3 293T Cell Overexpression Lysate (Denatured), Abnova
Supplier: Abnova Corporation H00004314T01
Description
Western Blotting
Specifications
293T | |
MMP3 293T Cell Transient Overexpression Lysate(Denatured) | |
54 | |
pCMV-MMP3 full-length | |
100 μL | |
Human |
Western Blot | |
NM_002422 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-MMP3 antibody (H00004314-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction