Learn More
A&D Engineering, Inc A&D Moisture Analyzers save valuable time and money by providing highly accurate results within minutes. Our SHS sensor technology provides moisture content even for the smallest sample sizes.
Supplier: A&D Engineering, Inc MF50
Save time and money with moisture results within minutes. MX models feature Super Hybrid Sensor SHS technology that gives ultra accurate moisture content even with small samples. Standard second radiant assist SRA feature allows for faster & uniform heating with halogen lamp, and our analyzers are simple to use and easy to maintain for all your loss-on-drying methods.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.