Learn More
Seca Corp 213 Portable stadiometer
SDP
Supplier:  Seca Corp 2131821009
Encompass_Preferred
At only 5.3 lbs, the seca 213 mobile stadiometer is especially suitable for use in the field but also ideal for doctors’ practices and pediatric hospitals. The measuring rod can be dismantled into several pieces and set up easily and quickly. The spacer keeps the rod straight and stable without any fittings. As the scale is printed along the side of the measuring rod, it is easy to read off the result while measuring, thus obtaining precise results – up to a height of 81 inches. • Simple and easy to set up – no wall fastening necessary • Large floor plate ensures stability • Includes heel positioner for ideal three-point measurement • The measuring rod can be dismantled and stored securely in the base for easy transport • Measuring range: 8 in – 6 ft 9 in / 20 – 205 cm • Graduation: 1/8 inch / 1 mm • Optional: carrying case seca 412
Catalog No. NC0242460
Explore available promotions