Learn More
Saint Gobain Performance Plastics TUB F4040A 5/16 x 7/16 50FT/PK
Supplier: Saint Gobain Performance Plastics AAG00022
Saint-Gobain Tygon Fuel and Lubricant Tubing Formula F-4040-A Diameter Outer 7/16 in. Maximum Pressure 25 psi Thickness 0.0625 in. Diameter Inner 5/16 in. Max. Operating Temperature 74 deg.C Min. Operating Temperature -37 deg.C Overall Length 50 ft. Yellow