Learn More
Saint Gobain Performance Plastics Tube Tygon 3/8 x 9/16 50FT-15M/PK
Supplier: Saint Gobain Performance Plastics ACF00028F
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Diameter Inner 3/8 in. Diameter Outer 9/16 in. Thickness 3/32 in. Overall Length 50 ft. 15 m Clear
This item is not returnable.
View return policy