Learn More
Stoelting™ Water Maze
Stoelting™ Water Maze is used to assess spatial and learning memory.
Catalog Number | Mfr. No. | Animal | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Animal | Price | Quantity & Availability | |||||
10-000-300
|
Stoelting™
60135 |
Mouse |
Each for $2,699.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
10-000-301
|
Stoelting™
60136 |
Mouse, Rat |
Each for $3,026.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
10-000-322
|
Stoelting™
60235 |
Rat |
Each for $3,199.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Water Maze tank is filled with water and equipped with a platform which acts as a means of escape for animals. During testing, animals learn the location of the platform using spatial cues.Specifications
Neuroscience |